MilliporeSigma™ Novagen™ B834(DE3) Competent Cells
Suitable for production of protein from target genes cloned in pET vectors by induction with IPTG
Supplier:  MilliporeSigma™ 690413
 View more versions of this product
Catalog No. 690413
Explore available promotions
Description
Description
  • B834 is parental strain for BL21 (1)
  • Protease-deficient hosts are methionine auxotrophs
  • Allows high specific-activity labeling of target proteins with methionine and selenomethionine for crystallography
Specifications
Specifications
≤ -70°C
Competent Cells
0.4mL