MilliporeSigma™ Novagen™ B834(DE3) Competent Cells
Suitable for production of protein from target genes cloned in pET vectors by induction with IPTG
$169.76 - $332.04
Description
- B834 is parental strain for BL21 (1)
- Protease-deficient hosts are methionine auxotrophs
- Allows high specific-activity labeling of target proteins with methionine and selenomethionine for crystallography
Specifications
≤ -70°C | |
Competent Cells |
For Research Use Only