missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Novagen™ CytoBuster™ Protein Extraction Reagent

Formulation of detergents optimized for efficient extraction of soluble proteins from mammalian and insect cells

$121.00 - $456.00

Specifications

Product Type Protein Extraction Reagent
For Use With (Application) Western blotting, immunoprecipitation and kinase/phosphatase assays
Content And Storage 15° to 30°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
710094MI
View Documents
MilliporeSigma™
MIL710094
250mL
Each for $456.00
 
710093MI
View Documents
MilliporeSigma™
MIL710093
50mL
Each for $121.00
 
Description

Description

  • Gentle, non-ionic composition of reagent enables isolation of functionally active endogenous or expressed proteins without need for secondary treatment such as sonication or freeze/thaw
  • Formulated for utilization in western blotting, immunoprecipitation and kinase/phosphatase assays
  • Reagent is compatible with protease inhibitors, kinase inhibitors and phosphatase inhibitors

Applications:

Western blotting, immunoprecipitation and kinase/phosphatase assays

Specifications

Specifications

Protein Extraction Reagent
Western blotting, immunoprecipitation and kinase/phosphatase assays
15° to 30°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'