missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ ELISA Stop Solution
A ready-to-use solution intended for use as a stop solution in ELISA applications that use horseradish peroxidase and TMB substrate solution.
$97.00 - $296.00
Specifications
Product Type | ELISA Stop Solution |
---|---|
Form | Liquid |
Substrate Type | HRP, TMB |
Content And Storage | Store at 2°C to 8°C |
Shipping Condition | Wet Ice |
Description
- A ready-to-use solution intended for use as a stop solution in ELISA applications that use horseradish peroxidase and TMB substrate solution
- As TMB substrate solution reacts with horseradish peroxidase, it produces a pronounced blue color
- At the end of TMB incubation, a volume of ELISA Stop Solution equal to the volume of TMB substrate solution is added to the wells to terminate the horseradish peroxidase reaction
- Addition of Stop Solution results in the rapid formation of a yellow end product
- The optical density of this yellow end product is measured at 450 nm
- The yellow end product is stable for sixty minutes after addition of Stop Solution, optical density should be measured within this time period
- Store at 2°C to 8°C
Order Info
Shipping Condition: Room Temperature
Specifications
ELISA Stop Solution | |
HRP, TMB | |
Wet Ice |
Liquid | |
Store at 2°C to 8°C |
For Research Use Only. Not for use in diagnostic procedures.