Invitrogen™ NuPAGE™ LDS Sample Buffer (4X) DFS Item
Used to prepare protein samples for denaturing gel electrophoresis with NuPAGE™ Novex™ gels. Invitrogen™ Novex™ NuPAGE™ LDS Sample Buffer (4X) contains lithium dodecyl sulfate, pH 8.4, which allows for maximum activity of the reducing agent.
Supplier:  Invitrogen™ NP0008
 View more versions of this product
Catalog No. NP0008
Explore available promotions
Description
Description
Optimal protein sample preparation for polyacrylamide gel electrophoresis (PAGE) requires denaturing and reducing protein disulfide bonds.
  • Highly viscous and concentrated solution that contains higher a concentration of glycerol
  • Contains twice the amount of LDS compared to the amount of SDS in Novex™ Tris-Glycine SDS Sample Buffer or in Tricine SDS Sample Buffer
  • NuPAGE™ LDS Sample Buffer should be brought to room temperature (25°C) prior to use
Specifications
Specifications
Sample Buffer (4X) containing lithium dodecyl sulfate (LDS) at pH 8.5 with SERVA Blue G250 and phenol red.
Store at 4°C to 25°C.
4X
LDS Sample Buffer
NuPAGE™
Approved for shipment at Room Temperature or on Wet Ice
Sample Loading Buffers
NuPAGE
NuPAGE™ Gels
250mL
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.