missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ NuPAGE™ LDS Sample Buffer (4X) DFS Item

Used to prepare protein samples for denaturing gel electrophoresis with NuPAGE™ Novex™ gels. Invitrogen™ Novex™ NuPAGE™ LDS Sample Buffer (4X) contains lithium dodecyl sulfate, pH 8.4, which allows for maximum activity of the reducing agent.

$38.62 - $262.00

Specifications

Content And Storage Sample Buffer (4X) containing lithium dodecyl sulfate (LDS) at pH 8.5 with SERVA Blue G250 and phenol red.

Store at 4°C to 25°C.

Buffer Sample Loading Buffers
Gel Type NuPAGE
Product Type LDS Sample Buffer
Gel Compatibility NuPAGE™ Gels
View More Specs

Includes: NuPAGE LDS Sample Buffer (4X), 10mL

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NP0007 DFS Item
View Documents
Invitrogen™
NP0007
10mL
Each for $38.62
 
NP0008 DFS Item
View Documents
Invitrogen™
NP0008
250mL
Each for $262.00
 
Description

Description

Use to prepare protein samples for denaturing gel electrophoresis with NuPAGE Novex™ gels. Optimal protein sample preparation for PAGE requires denaturing and reducing protein disulfide bonds.

  • Contains lithium dodecyl sulfate, pH 8.4, which allows for maximum activity of reducing agent
  • Highly viscous and concentrated solution
  • Contains higher concentration glycerol

NuPAGE LDS Sample Buffer should be brought to room temperature (25°C) prior to use.

1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Approved for shipment at Room Temperature or on Wet Ice

Specifications

Specifications

Sample Buffer (4X) containing lithium dodecyl sulfate (LDS) at pH 8.5 with SERVA Blue G250 and phenol red.

Store at 4°C to 25°C.

NuPAGE
NuPAGE™ Gels
Approved for shipment at Room Temperature or on Wet Ice
Sample Loading Buffers
LDS Sample Buffer
NuPAGE™
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.