Learn More
Nujol, for IR spectroscopy, Thermo Scientific Chemicals
For IR spectroscopy
$56.07 - $75.96
Chemical Identifiers
CAS | 8012-95-1 |
---|---|
Molecular Formula | MFCD00131611 |
Molecular Weight (g/mol) | 0.00 |
MDL Number | MFCD00131611 |
InChI Key | FFNDMZIBVDSQFI-UHFFFAOYSA-N |
Synonym | delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride |
PubChem CID | 68245 |
ChEBI | CHEBI:38701 |
IUPAC Name | 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride |
SMILES | * |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL14480AD
|
Thermo Scientific Chemicals
L14480AD |
50 mL |
Each for $56.07
|
|
|||||
AAL14480AP
|
Thermo Scientific Chemicals
L14480AP |
500 mL |
Each for $75.96
|
|
|||||
Description
Nujol is used for the synthetic resin used for cosmetics, fiber oil, food, medicine, as automotive lubricant, home decoration and in candle making.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNujol is used for the synthetic resin used for cosmetics, fiber oil, food, medicine, as automotive lubricant, home decoration and in candle making.
Solubility
Soluble in ether. Sparingly soluble in ethanol. Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and oxidizing agents.
Chemical Identifiers
8012-95-1 | |
0.00 | |
FFNDMZIBVDSQFI-UHFFFAOYSA-N | |
68245 | |
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride |
MFCD00131611 | |
MFCD00131611 | |
delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride | |
CHEBI:38701 | |
* |
Specifications
8012-95-1 | |
0.838 | |
220°C (428°F) | |
1.468 | |
50 mL | |
delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride | |
FFNDMZIBVDSQFI-UHFFFAOYSA-N | |
2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride | |
68245 | |
IR Spectroscopy |
∼-20°C | |
>300°C | |
MFCD00131611 | |
MFCD00131611 | |
14,7023 | |
Soluble in ether. Sparingly soluble in ethanol. Not miscible or difficult to mix in water. | |
* | |
0.00 | |
CHEBI:38701 | |
Nujol, for IR spectroscopy |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
EINECSNumber : 232-384-2
RTECSNumber : PY8030000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only