missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates

Store samples safely with these low-binding 96-well polypropylene plates, which will not allow DNA or proteins to bind and will allow for complete sample recovery.

Supplier:  Thermo Scientific™ 267342

 View more versions of this product

Catalog No. 12-565-370


Explore available promotions

Description

Description

Achieve complete sample recovery with Thermo ScientificTM NuncTM 96-Well Polypropylene MicroWellTM Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.

Features:

  • Shared-wall design increases the well volume
  • Wider wells and rounded bottom improve mixing
  • Conical bottom well for optimal sample recovery
  • Black and white plates available for fluorescent- and luminescent-based applications
  • Low binding polypropylene for homogeneous assays and storage
Specifications

Specifications

No Barcode
96-well
96-well Polypropylene Sample Processing & Storage Plates
Autoclavable
Low binding PP
Black
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
500 μL
450 μL
Non-treated
Yes
120
For Research Use Only. Not for use in diagnostic procedures.
Certified Quality
Certified RNase and DNase Free. ANSI Compliant.
Without Lid
8 x 12
10 pk., 120 cs.
Nunc
96-well plate
Polypropylene
96
Non-sterile
Round
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
10
Yes
ANSI/ SLAS Standard
Videos
SDS
Documents

Documents

Promotions

Promotions