missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates
Store samples safely with these low-binding 96-well polypropylene plates, which will not allow DNA or proteins to bind and will allow for complete sample recovery.
$953.53 - $2220.50
Specifications
Format | 96-well |
---|---|
Lid | Without Lid |
Description | 96-well Polypropylene Sample Processing & Storage Plates |
Array | 8 x 12 |
Binding Type | Low binding PP |
Catalog Number | Mfr. No. | Color | Well Shape | Sterility | Fit for Purpose | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Well Shape | Sterility | Fit for Purpose | Price | Quantity | ||
12-565-435
|
Thermo Scientific™
249943 |
Red | Round, Conical Bottom (V-well) | Non-sterile | Certified Quality |
Case of 120 Each for $1,320.45
|
|
||
12-565-436
|
Thermo Scientific™
249944 |
Natural | Round, Conical Bottom (V-well) | Non-sterile | Certified Quality |
Case of 120 Each for $1,284.38
|
|
||
12-565-437
|
Thermo Scientific™
249945 |
Black | Round, Conical Bottom (V-well) | Non-sterile | Certified Quality |
Case of 120 Each for $1,753.81
|
|
||
12-565-438
|
Thermo Scientific™
249946 |
Natural | Round, Conical Bottom (V-well) | Sterile | Sterile |
Case of 120 Each for $1,125.20
|
|
||
12-565-439
|
Thermo Scientific™
249947 |
Yellow | Round, Conical Bottom (V-well) | Non-sterile | Certified Quality |
Case of 120 Each for $1,376.34
|
|
||
12-565-440
|
Thermo Scientific™
249949 |
White | Round, Conical Bottom (V-well) | Non-sterile | Certified Quality |
Case of 120 Each for $2,220.50
|
|
||
12-565-441
|
Thermo Scientific™
249950 |
Blue | Round, Conical Bottom (V-well) | Non-sterile | Certified Quality |
Case of 120 Each for $1,363.93
|
|
||
12-565-368
|
Thermo Scientific™
267245 |
Natural | Round | Non-sterile | Certified Quality |
Case of 120 Each for $1,367.84
|
|
||
12-565-369
|
Thermo Scientific™
267334 |
Natural | Round | Sterile | Certified Quality |
Case of 120 Each for $1,643.13
|
|
||
12-565-370
|
Thermo Scientific™
267342 |
Black | Round | Non-sterile | Certified Quality |
Case of 120 Each for $1,873.63
|
|
||
12-565-371
|
Thermo Scientific™
267350 |
White | Round | Non-sterile | Certified Quality |
Case of 120 Each for $1,810.56
|
|
||
12-565-263
|
Thermo Scientific™
442587 |
Natural | Round, Conical Bottom (V-well) | Non-sterile | Certified Quality |
Case of 60 Each for $953.53
|
|
||
Description
Achieve complete sample recovery with Thermo ScientificTM NuncTM 96-Well Polypropylene MicroWellTM Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.
Features:
- Shared wall design increases the well volume
- Wider wells and rounded bottom improve mixing
- Conical bottom well for optimal sample recovery
- Black and white plates available for fluorescent- and luminescent-based applications
- Low binding polypropylene for homogeneous assays and storage
Specifications
96-well | |
96-well Polypropylene Sample Processing & Storage Plates | |
Low binding PP | |
96-well plate | |
Polypropylene | |
Non-treated | |
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
Yes |
Without Lid | |
8 x 12 | |
Nunc | |
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
96 | |
Yes | |
For Research Use Only. Not for use in diagnostic procedures. | |
ANSI/ SLAS Standard |