missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nunc™ 96-Well Polypropylene Sample Processing & Storage Microplates

Store samples safely with these low-binding 96-well polypropylene plates, which will not allow DNA or proteins to bind and will allow for complete sample recovery.

Supplier:  Thermo Scientific™ 249949

 View more versions of this product

Catalog No. 12-565-440


Explore available promotions

Description

Description

Achieve complete sample recovery with Thermo ScientificTM NuncTM 96-Well Polypropylene MicroWellTM Plates, which are the ideal storage plate as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from -80° to +121°C. Available in a variety of colors for quick identification of storage plates.

Features:

  • Shared-wall design increases the well volume
  • Wider wells and rounded bottom improve mixing
  • Conical bottom well for optimal sample recovery
  • Black and white plates available for fluorescent- and luminescent-based applications
  • Low binding polypropylene for homogeneous assays and storage
Specifications

Specifications

Custom Barcoding Available
96-well
96-well Polypropylene Sample Processing & Storage Plates
Autoclavable
Low binding PP
White
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
450 μL
400 μL
Non-treated
Yes
120
120/Cs.
Yes
ANSI/ SLAS Standard
Certified RNase and DNase Free. ANSI Compliant.
Without Lid
8 x 12
20 pk., 120 cs.
Nunc
96-well plate
Polypropylene
96
Non-sterile
Round, Conical Bottom (V-well)
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
20
For Research Use Only. Not for use in diagnostic procedures.
Certified Quality
SDS
Documents

Documents

Promotions

Promotions