Thermo Scientific™ Nunc™ Cryobank and Bank-It™ Replacement Caps
Ensure contamination-free cap replacement. Closures are supplied racked for manual or automated replacement.
Supplier:  Thermo Scientific™ 374021
 View more versions of this product
Catalog No. 37-402-1
Explore available promotions
Description
Description
Ensure contamination-free cap replacement with Thermo Scientific™ Nunc™ Cryobank and Bank-it™ Caps, which are supplied racked for manual or automated replacement. Available in natural, green, red or blue for simplified sample tracking.
Warranty: 90 days
  • Racked caps for automated or manual replacement of used caps
  • Racked in 96 microplate pattern for easy contamination free pick
Specifications
Specifications
Non-sterile
Closure
2D Barcode
Blue (Cap)
96
0.5mL and 1.0mL Cryobank and Bank-it Tubes
Blue
Polypropylene
Cryobank, Bank-It
10 racks of 96/case
960
Nunc™