missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nunc™ Microplate Lids

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.

Supplier:  Thermo Scientific™ 250005

 View more versions of this product

Catalog No. 12565462


Explore available promotions

Description

Description

Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.

  • Lids are for use with any standard plates
  • 96 Well Lids have individual condensation rings
  • Lids have notches for quick orientation
  • Universal Lids can be used on any standard height plate on the market
Specifications

Specifications

Clear
Polystyrene
384/1536
5 in.
No
180
5 x 3.35 in. (127 x 85 mm)
3.35 in.
Universal Lid
None
All well shapes
No
5 x 3.35 x 0.33 in. (127 x 85 x 8.34 mm)
Non-sterile
8.34 mm
127 mm
0.33 in.
20
No
85 mm
Nunc
-20°C to +80°C
All Microplates
Videos
SDS
Documents

Documents

Promotions

Promotions