Thermo Scientific™ Nunc™ Microplate Lids
Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.
$439.93 - $1274.00
Specifications
Material | Polystyrene |
---|---|
Length (English) | 5 in. |
Length (Metric) | 127 mm |
Width (English) | 3.35 in. |
Width (Metric) | 85 mm |
Catalog Number | Mfr. No. | Plate Compatibility | Sterility | Condensation Rings | Cut-off Corners | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Plate Compatibility | Sterility | Condensation Rings | Cut-off Corners | Price | Quantity | ||
12565476
|
Thermo Scientific™
250002 |
All Microplates | Sterile | No | No |
Case of 60 Each for $439.93
|
|
||
12565478
|
Thermo Scientific™
250003 |
All Microplates | Non-sterile | No | No |
Case of 60 Each for $561.94
|
|
||
12565462
|
Thermo Scientific™
250005 |
All Microplates | Non-sterile | No | No |
Case of 180 Each for $1,215.98
|
|
||
12565748
|
Thermo Scientific™
253623 |
All Microplates | Non-sterile | Yes | No |
Case of 100 Each for $708.91
|
|
||
12565749
|
Thermo Scientific™
253624 |
All Microplates | Sterile | Yes | No |
Case of 100 Each for $830.00
|
|
||
12-565-69
|
Thermo Scientific™
263339 |
96-well Plate | Non-sterile | Yes | Long Side |
Case of 100 Each for $751.55
|
|
||
12-565-70
|
Thermo Scientific™
264122 |
96-well Plate | Sterile | Yes | Long Side |
Case of 50 Each for $759.72
|
|
||
12-565-603
|
Thermo Scientific™
264611 |
384-well Plate | Sterile | No | Long Side |
Case of 60 Each for $552.50
|
|
||
12-565-604
|
Thermo Scientific™
264612 |
384-well Plate | Non-sterile | No | Long Side |
Case of 180 Each for $1,215.98
|
|
||
12-565-610
|
Thermo Scientific™
264616 |
384-well Plate | Sterile | No | Long Side |
Case of 180 Each for $1,274.00
|
|
||
12565477
|
Thermo Scientific™
264623 |
96-well Plate | Sterile | Yes | Long Side |
Case of 60 Each for $445.05
|
|
||
12-566-7
|
Thermo Scientific™
448143 |
384/1536-well Plate, OmniTray | Non-sterile | No | Long Side |
Case of 120 Each for $1,174.56
|
|
||
Description
Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.
- Lids are for use with any standard plates
- 96 Well Lids have individual condensation rings
- Lids have notches for quick orientation
- Universal Lids can be used on any standard height plate on the market
Specifications
Polystyrene | |
127 mm | |
85 mm |
5 in. | |
3.35 in. |