missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nunc™ Microplate Lids

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.

Supplier:  Thermo Scientific™ 263339

 View more versions of this product

Catalog No. 12-565-69


Explore available promotions

Description

Description

Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.

  • Lids are for use with any standard plates
  • 96 Well Lids have individual condensation rings
  • Lids have notches for quick orientation
  • Universal Lids can be used on any standard height plate on the market
Specifications

Specifications

Clear
Polystyrene
96
5 in.
Yes
100
5 x 3.35 in. (127 x 85 mm)
3.35 in.
Plate Lid
None
All well shapes
Long Side
5 x 3.35 x 0.3 in. (127 x 85 x 7.9 mm)
Non-sterile
7.9 mm
127 mm
0.3 in.
5
Yes
85 mm
Nunc
-20°C to +80°C
96-well Plate
Videos
SDS
Documents

Documents

Promotions

Promotions