AlphaProTech NuTech™ Coverall with Elastic Wrist and Ankle and Serged Seams
Strong, lightweight innovative microporous composite material protects the wearer from grime, dirt, and particulates.
Supplier: AlphaProTech CV640226
Description
- Protects from grime, dirt and particulates
- Pocketless to help control items entering and leaving work area
- Tapered collar neckline and a front zipper for easy donning and doffing
- With elastic wrist and ankle, serged seams
Specifications
Cleanroom Coveralls | |
Microporous Composite Fabric | |
Single-use | |
Collared | |
Non-sterile | |
NuTech |
2X-Large/3X-Large | |
White | |
Elastic | |
Serged | |
Controlled Environments |