missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Eisco™ Nylon Labware Brushes
Nylon Labware Brushes designed for cleaning a variety of labware feature nylon bristles and stainless steel handle to ensure durability.
$13.65 - $18.71
Specifications
Product Type | Cleaning Brush |
---|---|
Bristle Material | Nylon |
Bristle Type | Soft/Slightly Stiff |
Color-coded | Yes |
Handle Material | Stainless Steel Wire |
Description
- Fan shaped nylon bristle brush
- Stainless steel wire handle
- Ideal for cleaning a wide variety of glass and plastic labware
- Economically priced
Warranty
1 year limited manufacturer warranty.
Specifications
Cleaning Brush | |
Soft/Slightly Stiff | |
Stainless Steel Wire | |
Fan Tip | |
Non-sterile | |
Glassware and plasticware |
Nylon | |
Yes | |
Twisted Wire | |
Unscented | |
Cleaning labware | |
1 Ea. |