missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Phenylenediamine dihydrochloride, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 218480250

 View more versions of this product

Catalog No. AC218480250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

o-Phenylenediamine dihydrochloride
615-28-1
100.0
Pink to Purple
99+%
C6H8N2·2HCl
MFCD00012966
13, III, 28
RIIWUGSYXOBDMC-UHFFFAOYSA-N
benzene-1,2-diamine;dihydrochloride
11990
99+%
99+%
99.0
258°C
Authentic
Glass bottle
C6H4(NH2)2·2HCl
25 g
1,2-phenylenediamine dihydrochloride, o-phenylenediamine dihydrochloride, 1,2-benzenediamine, dihydrochloride, opd easy-tablets, opd tablets, benzene-1,2-diamine dihydrochloride, usaf ek-678, o-phenylenediamine.2hcl, 1,2-benzenediamine dihydrochloride, 1,2-diaminobenzene dihydrochloride
C1=CC=C(C(=C1)N)N.Cl.Cl
181.07
181.07
Crystalline Powder

Chemical Identifiers

615-28-1
181.07
RIIWUGSYXOBDMC-UHFFFAOYSA-N
11990
C1=CC=C(C(=C1)N)N.Cl.Cl
C6H8N2·2HCl
MFCD00012966
1,2-phenylenediamine dihydrochloride, o-phenylenediamine dihydrochloride, 1,2-benzenediamine, dihydrochloride, opd easy-tablets, opd tablets, benzene-1,2-diamine dihydrochloride, usaf ek-678, o-phenylenediamine.2hcl, 1,2-benzenediamine dihydrochloride, 1,2-diaminobenzene dihydrochloride
benzene-1,2-diamine;dihydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Toxic if swallowed.
Causes serious eye irritation.
Harmful if inhaled.<b

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the envi

GHS Signal Word: Danger

EINECSNumber : 210-418-7

RTECSNumber : ST0175000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only