missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Phenylenediamine dihydrochloride, 99+%, Thermo Scientific Chemicals

$329.70 - $965.69

Chemical Identifiers

CAS 615-28-1
Molecular Formula C6H8N2·2HCl
Molecular Weight (g/mol) 181.07
MDL Number MFCD00012966
InChI Key RIIWUGSYXOBDMC-UHFFFAOYSA-N
Synonym 1,2-phenylenediamine dihydrochloride, o-phenylenediamine dihydrochloride, 1,2-benzenediamine, dihydrochloride, opd easy-tablets, opd tablets, benzene-1,2-diamine dihydrochloride, usaf ek-678, o-phenylenediamine.2hcl, 1,2-benzenediamine dihydrochloride, 1,2-diaminobenzene dihydrochloride
PubChem CID 11990
IUPAC Name benzene-1,2-diamine;dihydrochloride
SMILES C1=CC=C(C(=C1)N)N.Cl.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC218480250
SDS View Documents
Thermo Scientific Chemicals
218480250
25 g Glass bottle
Each for $329.70
 
AC218481000
SDS View Documents
Thermo Scientific Chemicals
218481000
100 g Glass bottle
Each for $965.69
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

615-28-1
181.07
RIIWUGSYXOBDMC-UHFFFAOYSA-N
11990
C1=CC=C(C(=C1)N)N.Cl.Cl
C6H8N2·2HCl
MFCD00012966
1,2-phenylenediamine dihydrochloride, o-phenylenediamine dihydrochloride, 1,2-benzenediamine, dihydrochloride, opd easy-tablets, opd tablets, benzene-1,2-diamine dihydrochloride, usaf ek-678, o-phenylenediamine.2hcl, 1,2-benzenediamine dihydrochloride, 1,2-diaminobenzene dihydrochloride
benzene-1,2-diamine;dihydrochloride

Specifications

615-28-1
100.0
Pink to Purple
99+%
C6H8N2·2HCl
MFCD00012966
13, III, 28
RIIWUGSYXOBDMC-UHFFFAOYSA-N
benzene-1,2-diamine;dihydrochloride
11990
99+%
o-Phenylenediamine dihydrochloride, 99+%
99.0
258°C
Authentic
Glass bottle
C6H4(NH2)2·2HCl
25 g
1,2-phenylenediamine dihydrochloride, o-phenylenediamine dihydrochloride, 1,2-benzenediamine, dihydrochloride, opd easy-tablets, opd tablets, benzene-1,2-diamine dihydrochloride, usaf ek-678, o-phenylenediamine.2hcl, 1,2-benzenediamine dihydrochloride, 1,2-diaminobenzene dihydrochloride
C1=CC=C(C(=C1)N)N.Cl.Cl
181.07
181.07
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Toxic if swallowed.
Causes serious eye irritation.
Harmful if inhaled.<b

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the envi

GHS Signal Word: Danger

EINECSNumber : 210-418-7

RTECSNumber : ST0175000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only