Learn More
o-Phenylenediamine dihydrochloride, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 218481000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
o-Phenylenediamine dihydrochloride | |
615-28-1 | |
100.0 | |
Pink to Purple | |
99+% | |
C6H8N2·2HCl | |
MFCD00012966 | |
13, III, 28 | |
RIIWUGSYXOBDMC-UHFFFAOYSA-N | |
benzene-1,2-diamine;dihydrochloride | |
11990 | |
99+% |
99+% | |
99.0 | |
258°C | |
Authentic | |
Glass bottle | |
C6H4(NH2)2·2HCl | |
100 g | |
1,2-phenylenediamine dihydrochloride, o-phenylenediamine dihydrochloride, 1,2-benzenediamine, dihydrochloride, opd easy-tablets, opd tablets, benzene-1,2-diamine dihydrochloride, usaf ek-678, o-phenylenediamine.2hcl, 1,2-benzenediamine dihydrochloride, 1,2-diaminobenzene dihydrochloride | |
C1=CC=C(C(=C1)N)N.Cl.Cl | |
181.07 | |
181.07 | |
Crystalline Powder |
Chemical Identifiers
615-28-1 | |
181.07 | |
RIIWUGSYXOBDMC-UHFFFAOYSA-N | |
11990 | |
C1=CC=C(C(=C1)N)N.Cl.Cl |
C6H8N2·2HCl | |
MFCD00012966 | |
1,2-phenylenediamine dihydrochloride, o-phenylenediamine dihydrochloride, 1,2-benzenediamine, dihydrochloride, opd easy-tablets, opd tablets, benzene-1,2-diamine dihydrochloride, usaf ek-678, o-phenylenediamine.2hcl, 1,2-benzenediamine dihydrochloride, 1,2-diaminobenzene dihydrochloride | |
benzene-1,2-diamine;dihydrochloride |
Safety and Handling
GHS H Statement
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Toxic if swallowed.
Causes serious eye irritation.
Harmful if inhaled.<b
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the envi
GHS Signal Word: Danger
EINECSNumber : 210-418-7
RTECSNumber : ST0175000
TSCA : TSCA
RUO – Research Use Only