missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
o-Tolidine Dihydrochloride 98.0+%, TCI America™
$79.00 - $79.00
Chemical Identifiers
CAS | 612-82-8 |
---|---|
Molecular Formula | C14H18Cl2N2 |
Molecular Weight (g/mol) | 285.212 |
MDL Number | MFCD00012960 |
InChI Key | LUKPNZHXJRJBAN-UHFFFAOYSA-N |
Synonym | 3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl |
PubChem CID | 108938 |
IUPAC Name | 4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride |
SMILES | CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
T025425G
|
TCI America
T025425G |
25 g |
Each for $79.00
|
|
|||||
Chemical Identifiers
612-82-8 | |
285.212 | |
LUKPNZHXJRJBAN-UHFFFAOYSA-N | |
108938 | |
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl |
C14H18Cl2N2 | |
MFCD00012960 | |
3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl | |
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride |
Specifications
612-82-8 | |
MFCD00012960 | |
3077 | |
LUKPNZHXJRJBAN-UHFFFAOYSA-N | |
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride | |
108938 | |
≥98.0% (T) | |
o-Tolidine Dihydrochloride |
C14H18Cl2N2 | |
25 g | |
3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl | |
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl | |
285.212 | |
285.21 | |
Crystalline Powder |
Safety and Handling
RTECSNumber : DD1226000
TSCA : Yes