missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Tolidine Dihydrochloride 98.0+%, TCI America™

Supplier:  TCI America T025425G

 View more versions of this product

Catalog No. T025425G


Explore available promotions

Description

Description

Specifications

Specifications

o-Tolidine Dihydrochloride
C14H18Cl2N2
25 g
3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl
285.212
285.21
Crystalline Powder
612-82-8
MFCD00012960
3077
LUKPNZHXJRJBAN-UHFFFAOYSA-N
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
108938
≥98.0% (T)

Chemical Identifiers

612-82-8
285.212
LUKPNZHXJRJBAN-UHFFFAOYSA-N
108938
CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N)C)N.Cl.Cl
C14H18Cl2N2
MFCD00012960
3,3'-dimethylbenzidine dihydrochloride, o-tolidine dihydrochloride, 3,3'-dimethyl-1,1'-biphenyl-4,4'-diamine dihydrochloride, unii-5msk350kd8, dsstox_cid_511, dsstox_rid_75633, dsstox_gsid_20511, tolidine dihydrochloride, 1,1'-biphenyl-4,4'-diamine, 3,3'-dimethyl-, dihydrochloride, 3,3'-dimethylbenzidine.2hcl
4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
Videos
Safety and Handling

Safety and Handling

RTECSNumber : DD1226000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions