missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
o-Xylene, 99%, Extra Dry, AcroSeal™, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 443020010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
o-Xylene | |
0.8780g/mL | |
1L | |
1.5038 to 1.5058 | |
99% | |
C8H10 | |
MFCD00008519 | |
0.003% max. | |
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, miscible with ether and most organic solvents | |
CC1=CC=CC=C1C | |
106.17 | |
CHEBI:28063 | |
99% |
95-47-6 | |
143.0°C to 145.0°C | |
31°C | |
0.0003% max. | |
0.878 | |
C6H4(CH3)2 | |
0.81 mPa.s (20°C) | |
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes | |
CTQNGGLPUBDAKN-UHFFFAOYSA-N | |
1,2-xylene | |
7237 | |
106.17 | |
Extra Dry |
Chemical Identifiers
95-47-6 | |
106.17 | |
CTQNGGLPUBDAKN-UHFFFAOYSA-N | |
7237 | |
1,2-xylene |
C8H10 | |
MFCD00008519 | |
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes | |
CHEBI:28063 | |
CC1=CC=CC=C1C |
Safety and Handling
Danger
RUO â Research Use Only