missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Xylene, 99%, Extra Dry, AcroSeal™, Thermo Scientific™

Chemical Identifiers

CAS 95-47-6
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008519
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
Synonym o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
PubChem CID 7237
ChEBI CHEBI:28063
IUPAC Name 1,2-xylene
SMILES CC1=CC=CC=C1C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC443021000
SDS View Documents
Thermo Scientific Chemicals
443021000
100mL N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC443020010
SDS View Documents
Thermo Scientific Chemicals
443020010
1L N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

95-47-6
106.17
CTQNGGLPUBDAKN-UHFFFAOYSA-N
7237
1,2-xylene
C8H10
MFCD00008519
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
CHEBI:28063
CC1=CC=CC=C1C

Specifications

95-47-6
143.0°C to 145.0°C
31°C
0.0003% max.
0.878
C6H4(CH3)2
0.81 mPa.s (20°C)
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
CTQNGGLPUBDAKN-UHFFFAOYSA-N
1,2-xylene
7237
106.17
Extra Dry
0.8780g/mL
100mL
1.5038 to 1.5058
99%
C8H10
MFCD00008519
0.003% max.
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, miscible with ether and most organic solvents
CC1=CC=CC=C1C
106.17
CHEBI:28063
99%
o-Xylene
Safety and Handling

Safety and Handling

Danger

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only