missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

o-Xylene, 99%, Extra Dry, AcroSeal™, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 443021000

 View more versions of this product

Catalog No. AC443021000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

o-Xylene
0.8780g/mL
100mL
1.5038 to 1.5058
99%
C8H10
MFCD00008519
0.003% max.
Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, miscible with ether and most organic solvents
CC1=CC=CC=C1C
106.17
CHEBI:28063
99%
95-47-6
143.0°C to 145.0°C
31°C
0.0003% max.
0.878
C6H4(CH3)2
0.81 mPa.s (20°C)
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
CTQNGGLPUBDAKN-UHFFFAOYSA-N
1,2-xylene
7237
106.17
Extra Dry

Chemical Identifiers

95-47-6
106.17
CTQNGGLPUBDAKN-UHFFFAOYSA-N
7237
1,2-xylene
C8H10
MFCD00008519
o-xylene, 1,2-dimethylbenzene, ortho-xylene, o-xylol, o-methyltoluene, o-dimethylbenzene, 2-xylene, 3,4-xylene, benzene, 1,2-dimethyl, o-xylenes
CHEBI:28063
CC1=CC=CC=C1C
Safety and Handling

Safety and Handling

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only