pH Calibration Buffer Bottles, Oakton™
Use these PH buffers for meters with 0.01 resolution. Oakton™ pH Calibration Buffer Bottles are standardized against NIST-traceable pH references within 0.01 pH at 77°F (25°C).
Supplier:  Oakton Instruments 0065408
 View more versions of this product
Catalog No. 13-300-152
Explore available promotions
Description
Description
  • pH buffer solutions allow the user to adjust their system to read accurate measurements
  • Store buffers away from direct sunlight.
Specifications
Specifications
10.00
500 mL