missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

pH Calibration Buffer Bottles, Oakton™

Use these PH buffers for meters with 0.01 resolution. Oakton™ pH Calibration Buffer Bottles are standardized against NIST-traceable pH references within 0.01 pH at 77°F (25°C).

$41.91 - $66.44

Specifications

For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 1.68
Physical Form Liquid
Quantity 500 mL
Solution Type pH Buffer

Products 5
Catalog Number Mfr. No. pH Price Quantity  
Catalog Number Mfr. No. pH Price Quantity  
13-300-155
View Documents
Oakton Instruments
0065401
1.68
Each for $65.09
 
13-300-154
View Documents
Oakton Instruments
WD0065400
4.01
Each for $64.21
 
13-300-153
View Documents
Oakton Instruments
WD0065404
7.00
Each for $66.44
 
13-300-152
View Documents
Oakton Instruments
0065408
10.00
Each for $66.12
 
13-300-151
View Documents
Cole Parmer Instrument Co
0065412
12.46
Each for $41.91
 
Description

Description

  • pH buffer solutions allow the user to adjust their system to read accurate measurements
  • Store buffers away from direct sunlight.

Order Info

Shipping weight: 1-1/4 lbs. (0.6kg).

Certifications

NIST traceable, Certified to NIST traceable standards.

Specifications

Specifications

Calibration
1.68
500 mL
pH Meter and Electrode
Liquid
pH Buffer
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'