pH Calibration Buffer Bottles, Oakton™
Use these PH buffers for meters with 0.01 resolution. Oakton™ pH Calibration Buffer Bottles are standardized against NIST-traceable pH references within 0.01 pH at 77°F (25°C).
Supplier: Oakton Instruments WD0065400
Description
- pH buffer solutions allow the user to adjust their system to read accurate measurements
- Store buffers away from direct sunlight.
Specifications
4.01 | |
500 mL |