missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Octadecyltrichlorosilane, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 147401000

 View more versions of this product

Catalog No. AC147401000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Octadecyltrichlorosilane
94.0
Brown-Yellow to Colorless
223.0°C (10.0 mmHg)
Authentic
Glass bottle
1.4585 to 1.4605
MFCD00000484
0.98
Solubility in water: decomposes. Other solubilities: soluble in benzene, ethyl ether, heptane and, perchloroethylene
CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl
387.94
387.94
Liquid
112-04-9
100.0
0.9800g/mL
189°C
94% min. (GC)
C18H37Cl3Si
CH3(CH2)17SiCl3
100 mL
octadecyltrichlorosilane, trichloro octadecyl silane, n-octadecyltrichlorosilane, silane, trichlorooctadecyl, silane, octadecyltrichloro, n-octadecyl trichlorosilane, stearyltrichlorosilane, unii-1qle771pke, trichlorostearylsilane, trichloroctadecylsilane
PYJJCSYBSYXGQQ-UHFFFAOYSA-N
trichloro(octadecyl)silane
8157
95%

Chemical Identifiers

112-04-9
387.94
PYJJCSYBSYXGQQ-UHFFFAOYSA-N
8157
CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl
C18H37Cl3Si
MFCD00000484
octadecyltrichlorosilane, trichloro octadecyl silane, n-octadecyltrichlorosilane, silane, trichlorooctadecyl, silane, octadecyltrichloro, n-octadecyl trichlorosilane, stearyltrichlorosilane, unii-1qle771pke, trichlorostearylsilane, trichloroctadecylsilane
trichloro(octadecyl)silane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (or hair): Take off im

GHS Signal Word: Danger

EINECSNumber : 203-930-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only