Learn More
Octadecyltrichlorosilane, 95%, Thermo Scientific Chemicals
$84.73 - $617.81
Chemical Identifiers
CAS | 112-04-9 |
---|---|
Molecular Formula | C18H37Cl3Si |
Molecular Weight (g/mol) | 387.94 |
MDL Number | MFCD00000484 |
InChI Key | PYJJCSYBSYXGQQ-UHFFFAOYSA-N |
Synonym | octadecyltrichlorosilane, trichloro octadecyl silane, n-octadecyltrichlorosilane, silane, trichlorooctadecyl, silane, octadecyltrichloro, n-octadecyl trichlorosilane, stearyltrichlorosilane, unii-1qle771pke, trichlorostearylsilane, trichloroctadecylsilane |
PubChem CID | 8157 |
IUPAC Name | trichloro(octadecyl)silane |
SMILES | CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC147400250
|
Thermo Scientific Chemicals
147400250 |
25 mL | Glass bottle |
Each for $84.73
|
|
||||
AC147401000
|
Thermo Scientific Chemicals
147401000 |
100 mL | Glass bottle |
Each for $222.15
|
|
||||
AC147405000
|
Thermo Scientific Chemicals
147405000 |
500 mL | Glass bottle |
Each for $617.81
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-04-9 | |
387.94 | |
PYJJCSYBSYXGQQ-UHFFFAOYSA-N | |
8157 | |
CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl |
C18H37Cl3Si | |
MFCD00000484 | |
octadecyltrichlorosilane, trichloro octadecyl silane, n-octadecyltrichlorosilane, silane, trichlorooctadecyl, silane, octadecyltrichloro, n-octadecyl trichlorosilane, stearyltrichlorosilane, unii-1qle771pke, trichlorostearylsilane, trichloroctadecylsilane | |
trichloro(octadecyl)silane |
Specifications
112-04-9 | |
100.0 | |
0.9800g/mL | |
189°C | |
94% min. (GC) | |
C18H37Cl3Si | |
CH3(CH2)17SiCl3 | |
25 mL | |
octadecyltrichlorosilane, trichloro octadecyl silane, n-octadecyltrichlorosilane, silane, trichlorooctadecyl, silane, octadecyltrichloro, n-octadecyl trichlorosilane, stearyltrichlorosilane, unii-1qle771pke, trichlorostearylsilane, trichloroctadecylsilane | |
PYJJCSYBSYXGQQ-UHFFFAOYSA-N | |
trichloro(octadecyl)silane | |
8157 | |
95% | |
Octadecyltrichlorosilane |
94.0 | |
Brown-Yellow to Colorless | |
223.0°C (10.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4585 to 1.4605 | |
MFCD00000484 | |
0.98 | |
Solubility in water: decomposes. Other solubilities: soluble in benzene, ethyl ether, heptane and, perchloroethylene | |
CCCCCCCCCCCCCCCCCC[Si](Cl)(Cl)Cl | |
387.94 | |
387.94 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (or hair): Take off im
GHS Signal Word: Danger
EINECSNumber : 203-930-7
RUO – Research Use Only