missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Octanoic acid, 99%, Thermo Scientific Chemicals

Octanoic acid, 99%, C8H16O2, CAS Number-124-07-2, 1-heptanecarboxylic acid, caprylic acid, enantic acid, n-caprylic acid, neo-fat 8, n-octanoic acid, n-octoic acid, n-octylic acid, octoic acid, octylic acid, 1L, 385 to 393mg KOH/g, 98.5% min. (GC), 02, 347, 238.0 deg.C, 100.0, CHEBI:28837

Supplier:  Thermo Scientific Chemicals 129390010

 View more versions of this product

Catalog No. AC129390010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Octanoic acid
385 to 393mg KOH/g
100.0
Colorless to Yellow
238.0°C
Authentic
Glass bottle
1.4268 to 1.4288
MFCD00004429
02, 347
15, 1767
Solubility in water: 0.68g/L (20°C). Other solubilities: soluble in alcohol and ether
CCCCCCCC(=O)O
144.21
5.8 mPa.s (20°C)
144.21
Liquid
124-07-2
98.5
16.0°C to 16.5°C
0.9100g/mL
110°C
98.5% min. (GC)
C8H16O2
CH3(CH2)6CO2H
1 L
0.91
caprylic acid, n-octanoic acid, octylic acid, n-caprylic acid, octoic acid, n-octylic acid, n-octoic acid, neo-fat 8, 1-heptanecarboxylic acid, enantic acid
WWZKQHOCKIZLMA-UHFFFAOYSA-N
octanoic acid
379
CHEBI:28837
99%
ACROS Organics

Chemical Identifiers

124-07-2
144.21
WWZKQHOCKIZLMA-UHFFFAOYSA-N
379
octanoic acid
C8H16O2
MFCD00004429
caprylic acid, n-octanoic acid, octylic acid, n-caprylic acid, octoic acid, n-octylic acid, n-octoic acid, neo-fat 8, 1-heptanecarboxylic acid, enantic acid
CHEBI:28837
CCCCCCCC(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 204-677-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only