Learn More
Octanoic acid, 99%
Octanoic acid, 99%, C8H16O2, CAS Number-124-07-2, 1-heptanecarboxylic acid, caprylic acid, enantic acid, n-caprylic acid, neo-fat 8, n-octanoic acid, n-octoic acid, n-octylic acid, octoic acid, octylic acid, 1L, 385 to 393mg KOH/g, 98.5% min. (GC), 02, 347, 238.0 deg.C, 100.0, CHEBI:28837 | CAS: 124-07-2 | C8H16O2 | 144.21 g/mol
$88.42 - $1106.78
Chemical Identifiers
| CAS | 124-07-2 |
|---|---|
| Molecular Formula | C8H16O2 |
| Molecular Weight (g/mol) | 144.21 |
| MDL Number | MFCD00004429 |
| InChI Key | WWZKQHOCKIZLMA-UHFFFAOYSA-N |
| Synonym | caprylic acid, n-octanoic acid, octylic acid, n-caprylic acid, octoic acid, n-octylic acid, n-octoic acid, neo-fat 8, 1-heptanecarboxylic acid, enantic acid |
| PubChem CID | 379 |
| ChEBI | CHEBI:28837 |
| IUPAC Name | octanoic acid |
| SMILES | CCCCCCCC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC129390010
|
Thermo Scientific Chemicals
129390010 |
1 L | Glass bottle |
Each for $88.42
|
|
||||
|
AC129390025
|
Thermo Scientific Chemicals
129390025 |
2.5 L | Glass bottle |
Each for $167.73
|
|
||||
|
AC129390251
|
Thermo Scientific Chemicals
129390251 |
25 L | Metal drum |
Each for $1,106.78
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 124-07-2 | |
| 144.21 | |
| WWZKQHOCKIZLMA-UHFFFAOYSA-N | |
| 379 | |
| octanoic acid |
| C8H16O2 | |
| MFCD00004429 | |
| caprylic acid, n-octanoic acid, octylic acid, n-caprylic acid, octoic acid, n-octylic acid, n-octoic acid, neo-fat 8, 1-heptanecarboxylic acid, enantic acid | |
| CHEBI:28837 | |
| CCCCCCCC(=O)O |
Specifications
| 124-07-2 | |
| 98.5 | |
| 16.0°C to 16.5°C | |
| 0.9100g/mL | |
| 110°C | |
| 98.5% min. (GC) | |
| C8H16O2 | |
| CH3(CH2)6CO2H | |
| 1 L | |
| 0.91 | |
| caprylic acid, n-octanoic acid, octylic acid, n-caprylic acid, octoic acid, n-octylic acid, n-octoic acid, neo-fat 8, 1-heptanecarboxylic acid, enantic acid | |
| WWZKQHOCKIZLMA-UHFFFAOYSA-N | |
| octanoic acid | |
| 379 | |
| CHEBI:28837 | |
| 99% | |
| ACROS Organics |
| 385 to 393mg KOH/g | |
| 100.0 | |
| Colorless to Yellow | |
| 238.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4268 to 1.4288 | |
| MFCD00004429 | |
| 02, 347 | |
| 15, 1767 | |
| Solubility in water: 0.68g/L (20°C). Other solubilities: soluble in alcohol and ether | |
| CCCCCCCC(=O)O | |
| 144.21 | |
| 5.8 mPa.s (20°C) | |
| 144.21 | |
| Liquid | |
| Octanoic acid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 204-677-5
RUO – Research Use Only