missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 416500250

 View more versions of this product

Catalog No. AC416500250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ion pair chromatography
Specifications

Specifications

Octyl sulfate, sodium salt
(5% aq. soln.) (1cm cell), (5% aq. soln.) (1 cm cell) at 0nm, 0.02 max. at 250nm
White
Authentic
Glass bottle
CH3(CH2)7OSO3Na
sodium octyl sulfate, sodium n-octyl sulfate, sodium octyl sulphate, sipex ols, cycloryl os, duponol 80, sodium capryl sulfate, octyl sodium sulfate, sulfuric acid, monooctyl ester, sodium salt, octyl sulfate sodium salt
WFRKJMRGXGWHBM-UHFFFAOYSA-M
sodium;octyl sulfate
2735107
99%
142-31-4
210.0°C
2% (105°C) (vacuum/P2O5) max.
1% max. Alkyl sulphate homologues (GC)
C8H15Na·H2SO4
25 g
(25% in water at 25°C) Clear colorless
CCCCCCCCOS(=O)(=O)[O-].[Na+]
232.28
232.28
Coarse Powder

Chemical Identifiers

142-31-4
232.28
sodium octyl sulfate, sodium n-octyl sulfate, sodium octyl sulphate, sipex ols, cycloryl os, duponol 80, sodium capryl sulfate, octyl sodium sulfate, sulfuric acid, monooctyl ester, sodium salt, octyl sulfate sodium salt
sodium;octyl sulfate
C8H15Na·H2SO4
WFRKJMRGXGWHBM-UHFFFAOYSA-M
2735107
CCCCCCCCOS(=O)(=O)[O-].[Na+]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Danger

EINECSNumber : 205-535-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only