Learn More
Octyl sulfate, sodium salt, 99%, HPLC grade, Thermo Scientific Chemicals
$1379.47 - $4552.33
Chemical Identifiers
CAS | 142-31-4 |
---|---|
Molecular Formula | C8H15Na·H2SO4 |
Molecular Weight (g/mol) | 232.28 |
InChI Key | WFRKJMRGXGWHBM-UHFFFAOYSA-M |
Synonym | sodium octyl sulfate, sodium n-octyl sulfate, sodium octyl sulphate, sipex ols, cycloryl os, duponol 80, sodium capryl sulfate, octyl sodium sulfate, sulfuric acid, monooctyl ester, sodium salt, octyl sulfate sodium salt |
PubChem CID | 2735107 |
IUPAC Name | sodium;octyl sulfate |
SMILES | CCCCCCCCOS(=O)(=O)[O-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC416500250
|
Thermo Scientific Chemicals
416500250 |
25 g | Glass bottle |
Each for $1,379.47
|
|
||||
AC416501000
|
Thermo Scientific Chemicals
416501000 |
100 g | Glass bottle |
Each for $4,552.33
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ion pair chromatographyChemical Identifiers
142-31-4 | |
232.28 | |
sodium octyl sulfate, sodium n-octyl sulfate, sodium octyl sulphate, sipex ols, cycloryl os, duponol 80, sodium capryl sulfate, octyl sodium sulfate, sulfuric acid, monooctyl ester, sodium salt, octyl sulfate sodium salt | |
sodium;octyl sulfate |
C8H15Na·H2SO4 | |
WFRKJMRGXGWHBM-UHFFFAOYSA-M | |
2735107 | |
CCCCCCCCOS(=O)(=O)[O-].[Na+] |
Specifications
142-31-4 | |
210.0°C | |
2% (105°C) (vacuum/P2O5) max. | |
1% max. Alkyl sulphate homologues (GC) | |
C8H15Na·H2SO4 | |
25 g | |
(25% in water at 25°C) Clear colorless | |
CCCCCCCCOS(=O)(=O)[O-].[Na+] | |
232.28 | |
232.28 | |
Coarse Powder |
(5% aq. soln.) (1cm cell), (5% aq. soln.) (1 cm cell) at 0nm, 0.02 max. at 250nm | |
White | |
Authentic | |
Glass bottle | |
CH3(CH2)7OSO3Na | |
sodium octyl sulfate, sodium n-octyl sulfate, sodium octyl sulphate, sipex ols, cycloryl os, duponol 80, sodium capryl sulfate, octyl sodium sulfate, sulfuric acid, monooctyl ester, sodium salt, octyl sulfate sodium salt | |
WFRKJMRGXGWHBM-UHFFFAOYSA-M | |
sodium;octyl sulfate | |
2735107 | |
99% | |
Octyl sulfate, sodium salt |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
GHS Signal Word: Danger
EINECSNumber : 205-535-5
RUO – Research Use Only