missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OHAUS™ Ranger™ 7000 Compact Scales

Get the weighing scale with the highest resolution, largest display and largest memory library of any industrial bench scale in the Ohaus portfolio. The Ohaus Ranger™ 7000 Compact Scale has a sturdy industrial and modular design that supports flexible and heavy-duty use.

Supplier:  OHAUS™ 30070311

 View more versions of this product

Catalog No. 01-920-126


Explore available promotions

Description

Description

  • Built with IP54 cast metal housing and sealed metal indicator that ensure durability
  • Sturdy industrial and modular design supports flexible and Heavy-Duty use
  • Metal indicator and housing allows long time operation in rugged and harsh industrial environments
  • Scale with 10 advanced application modes for variety of complex applications that minimize need for manual calculations and control peripheral devices
  • Bright backlit, 4.3 in. graphic TFT display can be operated in multiple languages
  • Navigation panel comprised of keypad with 5 quick, 8 function and 12 alphanumeric keys
  • SmarText™ 2.0 Software enabled scale is simple to use with minimum training for users
  • Precise display resolution up to 75000d
  • Weighing units: kg, g, lb, oz, lb:oz
  • Quick results with one second stabilization time
  • Fast performance and Legal for Trade Certifications assures accurate results
  • Data produced can be easily exported through standard RS232 or USB device port
  • Additional connectivity options include second RS232 or Ethernet, and standard USB host port for bar code scanner or flash drive
  • With full GLP/GMP data output, Ranger 7000 scale can help meet traceability and compliance requirements
Application Modes
  • Weighing
  • Parts Counting
  • Percent Weighing
  • Check Weighing
  • Dynamic Weighing (Display Hold)
  • Filling
  • Formulation
  • Differential Weighing
  • Density Weighing
Specifications

Specifications

Stainless Steel Platform, Die Cast Housing
LCD
IP54
1 sec.
100/240 VAC 50/60 Hz Universal Power Supply, rechargeable lithium battery (Optional)
15 kg
0.2 g (0.0005 lb.)
2% or 10% of Capacity
377 x 311 mm
Class III
Videos
SDS
Documents

Documents

Promotions

Promotions