missing translation for 'onlineSavingsMsg'
Learn More
S'il vous plaît vous connecter à votre compte en ligne pour afficher votre prix réduits

OHAUS™ Ranger™ 7000 Compact Scales

Get the weighing scale with the highest resolution, largest display and largest memory library of any industrial bench scale in the Ohaus portfolio. The Ohaus Ranger™ 7000 Compact Scale has a sturdy industrial and modular design that supports flexible and heavy-duty use.

$3784.48 - $10539.30

Spécifications

Construction Stainless Steel Platform, Die Cast Housing
Display Type LCD
Protection Class IP54
Range 2% or 10% of Capacity
Stabilization Time 1 sec.
voir les spécifications

Produits 8
Numéro de catalogue Numéro du manufacturier. Capacity Readability Dimensions (L x W x H) Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Capacity Readability Dimensions (L x W x H) Prix Quantité  
01-920-120
Afficher les documents
OHAUS™
30088840
3 kg 0.01 g (0.0002 lb.) 240 x 240 mm
chaque for $5,944.70
 
01-920-124
Afficher les documents
OHAUS™
30070289
3 kg 0.05 g (0.0001 lb.) 280 x 280 mm
chaque for $3,784.48
 
01-920-123
Afficher les documents
OHAUS™
30088843
35 kg 0.1 g (0.0002 lb.) 377 x 311 mm N/A
N/A
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
01-920-122
Afficher les documents
OHAUS™
30088842
15 kg 0.1 g (0.0002 lb.) 377 x 311 mm
chaque for $8,744.66
 
01-920-121
Afficher les documents
OHAUS™
30088841
6 kg 0.02 g (0.00005 lb.) 240 x 240 mm
chaque for $6,616.11
 
01-920-125
Afficher les documents
OHAUS™
30070310
6 kg 0.1 g (0.0002 lb.) 280 x 280 mm
chaque for $3,784.48
 
01-920-126
Afficher les documents
OHAUS™
30070311
15 kg 0.2 g (0.0005 lb.) 377 x 311 mm
chaque for $3,784.48
 
01-920-127
Afficher les documents
OHAUS™
30070312
35 kg 0.5 g (0.0011 lb.) 377 x 311 mm
chaque for $3,784.48
 
Description

Description

 

  • Built with IP54 cast metal housing and sealed metal indicator that ensure durability
  • Metal indicator and housing allows long time operation in rugged and harsh industrial environments
  • Scale with 10 advanced application modes for variety of complex applications that minimize need for manual calculations and control peripheral devices
  • Bright backlit, 4.3 in. graphic TFT display can be operated in multiple languages
  • Navigation panel comprised of keypad with 5 quick, 8 function and 12 alphanumeric keys
  • SmarText™ 2.0 Software enabled scale is simple to use with minimum training for users
  • Precise display resolution up to 75000d
  • Weighing units: kg, g, lb, oz, lb:oz
  • Quick results with one second stabilization time
  • Fast performance and Legal for Trade Certifications assures accurate results
  • Data produced can be easily exported through standard RS232 or USB device port
  • Additional connectivity options include second RS232 or Ethernet, and standard USB host port for bar code scanner or flash drive
  • With full GLP/GMP data output, Ranger 7000 scale can help meet traceability and compliance requirements

Application Modes
  • Weighing
  • Parts Counting
  • Percent Weighing
  • Check Weighing
  • Dynamic Weighing (Display Hold)
  • Filling
  • Formulation
  • Differential Weighing
  • Density Weighing

 

Spécifications

Spécifications

Stainless Steel Platform, Die Cast Housing
IP54
1 sec.
LCD
2% or 10% of Capacity
100/240 VAC 50/60 Hz Universal Power Supply, rechargeable lithium battery (Optional)
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'