Invitrogen™ OneComp eBeads™ Compensation Beads
Spherical particles that can be stained with individual fluorochrome-conjugated antibodies for use as single-color compensation controls. OneComp eBeads react with antibodies of mouse, rat and hamster origin, and are immunoglobulin light chain-independent. 1 drop (50μL)/test
Supplier:  Invitrogen™ 01111142
 View more versions of this product
Catalog No. 501129031
Explore available promotions
Alert:
Contains Sodium azide
Description
Description
  • Each drop of beads contains both a positive population that will capture any mouse, rat or hamster antibody and a negative population that will not react with antibody
  • This bimodal distribution can be used for single-color compensation controls in multicolor flow cytometry experiments
  • Cross-reacts to some antibodies of rabbit origin, but have not been extensively tested for this reactivity
  • Designed for use in compensation with all fluorochromes excited by blue (488nm), green (532nm), yellow-green (561nm), and red (633 to 635nm) lasers
  • Compatible with eFluor™ 450 but is not optimized for compensation of other fluorochromes excited by a violet (405nm) laser
Specifications
Specifications
100 Tests
100 tests
Flow Cytometer
Compensation Beads
eBeads™
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.