missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ OneComp eBeads™ Compensation Beads
Spherical particles that can be stained with individual fluorochrome-conjugated antibodies for use as single-color compensation controls. OneComp eBeads react with antibodies of mouse, rat and hamster origin, and are immunoglobulin light chain-independent. 1 drop (50μL)/test
$172.00 - $414.00
Specifications
Product Line | eBeads™ |
---|---|
Product Type | Compensation Beads |
For Use With (Equipment) | Flow Cytometer |
Alert:
Contains Sodium azide
Description
- Each drop of beads contains both a positive population that will capture any mouse, rat or hamster antibody and a negative population that will not react with antibody
- This bimodal distribution can be used for single-color compensation controls in multicolor flow cytometry experiments
- Cross-reacts to some antibodies of rabbit origin, but have not been extensively tested for this reactivity
- Designed for use in compensation with all fluorochromes excited by blue (488nm), green (532nm), yellow-green (561nm), and red (633 to 635nm) lasers
- Compatible with eFluor™ 450 but is not optimized for compensation of other fluorochromes excited by a violet (405nm) laser
Specifications
eBeads™ | |
Compensation Beads | |
Flow Cytometer |
For Research Use Only. Not for use in diagnostic procedures.