Invitrogen™ OneComp eBeads™ Compensation Beads
Spherical particles that can be stained with individual fluorochrome-conjugated antibodies for use as single-color compensation controls. OneComp eBeads react with antibodies of mouse, rat and hamster origin, and are immunoglobulin light chain-independent. 1 drop (50μL)/test
Supplier: Invitrogen™ 01111141
Alert:
Contains Sodium azide
Description
- Each drop of beads contains both a positive population that will capture any mouse, rat or hamster antibody and a negative population that will not react with antibody
- This bimodal distribution can be used for single-color compensation controls in multicolor flow cytometry experiments
- Cross-reacts to some antibodies of rabbit origin, but have not been extensively tested for this reactivity
- Designed for use in compensation with all fluorochromes excited by blue (488nm), green (532nm), yellow-green (561nm), and red (633 to 635nm) lasers
- Compatible with eFluor™ 450 but is not optimized for compensation of other fluorochromes excited by a violet (405nm) laser
Specifications
25 tests | |
25 tests | |
Flow Cytometer |
Compensation Beads | |
eBeads™ |
For Research Use Only. Not for use in diagnostic procedures.