Simport™ Scientific Capinsert™ For Cryovial Tubes
The Capinsert™ For Cryovial Tubes inserts fit precisely into the cap of the Cryovial™ for color identification.
Supplier:  Simport™ Scientific T31211
 View more versions of this product
Catalog No. 22-038-503
Explore available promotions
Description
Description
  • Color coded inserts
  • Made of polypropylene
  • For Cryovial tubes
Specifications
Specifications
Burnt Orange
Polypropylene
Cryovial™ Tubes
Product Suggestions
Product Suggestions