missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Orion™ Single Junction Refillable Epoxy Bodied pH/ATC Triode™
Accurately measure pH and temperature together in just one electrode, with a built-in temperature sensor.
$1263.94 - $1273.89
Specifications
Junction | Glass Fiber |
---|---|
Material | Epoxy |
pH Range | 0 to 14 |
Temperature (Metric) Operating | 0°C to 90°C |
For Use With (Application) | ideal for field or plant use. |
Catalog Number | Mfr. No. | Connector Types | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Connector Types | Price | Quantity | ||||||
13-641-259
|
Thermo Scientific™
9157BN |
BNC |
Each for $1,263.94
|
|
||||||
13-642-252
|
Thermo Scientific™
9157BNMD |
Mini DIN |
Each for $1,273.89
|
|
||||||
Description
Thermo Scientific™ Refillable Orion™ Triode™ 3-in-1 pH/Automatic Temperature Compensation Electrodes have an built-in ATC. They are engineered with an epoxy body for ruggedness and best suited for general purpose samples.
Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.
Solutions required
Electrode fill solution
900011 4M KCl with Ag/AgCl filling solution, 5 x 60 mL bottles
Electrode storage solution
910001 pH electrode storage solution, 475 mL bottle
pH buffers
910168 pH 1.68 buffer, NIST traceable, 475 mL
910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112 pH 12.46 buffer, NIST traceable, 475 mL
910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
electrode storage solution, 475ml, and 12 mm electrode storage bottle
electrode storage solution, 475ml, and 12 mm electrode storage bottle
pH cleaning solution kit
900020 Cleaning solution A and C, 30 ml each. B and D, 60 ml each
Items included with
900011 4M KCl with Ag/AgCl filling solution 60mL 1 bottle
Warranty period
12 months from date of purchase
Specifications
Glass Fiber | |
0 to 14 | |
ideal for field or plant use. | |
Ag/AgCl | |
4.7 in. | |
0.47 in. | |
16 mm |
Epoxy | |
0°C to 90°C | |
Orion pH Meter (field or plant-use) | |
32°F to 194°F | |
120 mm | |
12 mm |