missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Orion™ Nitrate Electrodes
Measurements are quick, simple, accurate and economical. Analyze free nitrate ions in aqueous solutions reliably at low limits of detection.
Supplier: Thermo Scientific™ 9707BNWP
Description
Measure nitrate in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Nitrate Electrodes.
- Calibration standards catalog number: 920706 — 0.1M NaNO3; 920707 — 1000ppm N; 930707 — 100ppm N
- Required ISA catalog number: 930711 or 930710/nitrate ISS
Specifications
BNC | |
drift-free potentials | |
ISE Meters |
Combination Nitrate Electrode with Waterproof BNC Connector | |
Free-flowing Sure-Flow™ junction assures stable | |
Combination Ion Selective Electrode |