Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions
Restore proper electrode performance and prolong electrode life using these pH electrode cleaning solutions.
Supplier:  Thermo Scientific™ 900020
 View more versions of this product
Catalog No. 13-642-583
Explore available promotions
Description
Description
Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions in ready to use bottles.
Cleaning Solutions:
  • Solutions are identified by the letters A, B, C, and D; each letter designates a unique formulation
  • Cleaning solution A is for removing protein contaminants, cleaning solution B is for remov
Specifications
Specifications
1 Bottle Each of Cleaning Solutions A, B, C, and D; Beaker and Pipet
pH Electrode Cleaning Solution
See individual components
pH Electrode
Solutions restore proper electrode performance and prolong the useful life of the electrode
See individual components
Videos
SDS
Documents
Documents
Promotions
Promotions
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.