Learn More
Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions
Restore proper electrode performance and prolong electrode life using these pH electrode cleaning solutions.
$157.18 - $256.97
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
13-642-583
|
Thermo Scientific™
900020 |
1 Bottle Each of Cleaning Solutions A, B, C, and D; Beaker and Pipet |
Pack of 4 Each for $256.97
|
|
|||||
13-642-584
|
Thermo Scientific™
900021WA |
4 x 1 oz. (4 x 30mL) Bottles of Cleaning Solution A for Protein Removal, Beaker and Pipet |
Pack of 4 Each for $172.74
|
|
|||||
13-642-585
|
Thermo Scientific™
900022WA |
4 x 2 oz. (4 x 60mL) Bottles of Cleaning Solution B for Bacteria Removal, Beaker and Pipet |
Pack of 4 Each for $181.15
|
|
|||||
13-642-586
|
Thermo Scientific™
900023 |
4 x 1 oz. (4 x 30mL) Bottles of Cleaning Solution C for General Cleaning, Beaker and Pipet |
Pack of 4 Each for $176.07
|
|
|||||
13-642-587
|
Thermo Scientific™
ORI900024 |
Solution D 4 x 2 oz. (4 x 60mL) Bottles of Cleaning Solution D for Oil and Grease Removal, Beaker and Pipet |
Pack of 4 Each for $176.07
|
|
|||||
13-641-273
|
Thermo Scientific™
911110 |
10 Pack of pH Electrode Rinse Solution Pouches |
Pack of 10 Each for $157.18
|
|
|||||
13-641-277
|
Thermo Scientific™
911125 |
25 Pack of pH Electrode Rinse Solution Pouches |
Pack of 25 Each for $237.85
|
|
|||||
Description
Restore proper electrode performance and prolong electrode life using Thermo Scientific™ Orion™ pH Electrode Cleaning Solutions in ready to use bottles.
Cleaning Solutions:
- Solutions are identified by the letters A, B, C, and D; each letter designates a unique formulation
- Cleaning solution A is for removing protein contaminants, cleaning solution B is for removing bacterial contaminants, cleaning solution C is for general cleaning and cleaning solution D is for removing oil and grease contaminants
- Bottles include a small beaker to hold the solution and a plastic pipet to remove the old electrode filling solution
Includes (PN 900020):
- One bottle each of cleaning solutions A, B, C and D
- One beaker
- One pipet
Note:
The following part numbers have been changed due to a recent system upgrade:
Old Part Number: 900021
New Part Number: 900021-WA
Old Part Number: 900022
New Part Number: 900022-WA
Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.
Warranty: Out of box
Specifications
pH Electrode |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.