Thermo Scientific™ Orion™ pH Electrode Filling Solution Edge
Edge
Use this pH electrode filling solution with pH electrodes and reference electrodes.
Supplier:  Thermo Scientific™ ORI810007
 View more versions of this product
Catalog No. 13-641-755
Edge
Explore available promotions
Description
Description
Use Thermo Scientific™ Orion™ pH Electrode Filling Solution with pH electrodes and reference electrodes. Packaged as five 2 oz. (5 x 60mL) bottles.
  • Only use ROSS™ filling solution (810007) with ROSS™ and ROSS Ultra™ pH electrodes to prevent damaging the ROSS electrodes
  • When the single junction reference electrode (900100) or double junction reference electrode (900200) are used with Orion ion selective electrodes (ISE), refer to the recommended ISE filling solution listed in the ISE user guide
Specifications
Specifications
Filling Solution for ROSS Ultra, ROSS, PerpHecT ROSS Electrodes
pH Electrode Filling Solution
5 2 oz. (5 x 60 mL) Bottles
pH Electrodes and Reference Electrodes (To prevent damage, only use ROSS filling solution with Ross Electrodes)
Surfactant Electrode Fill, Ross Fill
60 mL
Videos
SDS
Documents
Documents
Promotions
Promotions
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.