Thermo Scientific™ Orion™ pH Electrode Filling Solution
Use this pH electrode filling solution with pH electrodes and reference electrodes.
$253.76 - $366.96
Specifications
Type pH Electrode Filling Solution
Quantity 60 mL
View More Specs
Products 9
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
13-642-402
View Documents Promotion Details
Thermo Scientific™
ORI510011
Filling Solution for No Cal pH Electrodes
Each for $366.96
13-641-755 Edge
View Documents Promotion Details
Thermo Scientific™
ORI810007
Filling Solution for ROSS Ultra, ROSS, PerpHecT ROSS Electrodes
Pack of 5 Each for $274.91
Edge
13-641-899
View Documents Promotion Details
Thermo Scientific™
ORI900001
Filling Solution for Single Junction Sure-Flow Reference Electrodes
Pack of 5 Each for $265.30
13-641-901
View Documents Promotion Details
Thermo Scientific™
900002
Double Junction Sure-Flow Reference Electrode, Inner Chamber
Pack of 5 Each for $278.76
13-641-897
View Documents Promotion Details
Thermo Scientific™
900003
Double Junction Sure-Flow Reference Electrode, Outer Chamber
Pack of 5 Each for $278.76
13-641-685
View Documents Promotion Details
Thermo Scientific™
900004
Standard, Specialty, PerpHecT Ag/AgCl and Single Junction Refillable Green pH Electrodes Used in Low Ionic Strength Samples
Pack of 5 Each for $280.67
13-641-686
View Documents Promotion Details
Thermo Scientific™
900011
Standard, Specialty, PerpHecT Ag/AgCl, Single Junction Refillable Green and Micro pH Electrodes, ORP Electrodes
Pack of 5 Each for $253.76
13-641-845
View Documents Promotion Details
Thermo Scientific™
ORI900019
Steam Sterilizable Electrodes
Each for $365.13
13-200-312
View Documents Promotion Details
Thermo Scientific™
ORI910008WA
Filling Solution for Double Junction, Double Junction KNIpHE™ Electrodes
Case of 5 Each for $257.43
Description
Description
Use Thermo Scientific™ Orion™ pH Electrode Filling Solution with pH electrodes and reference electrodes. Packaged as five 2 oz. (5 x 60mL) bottles.
  • Only use ROSS™ filling solution (810007) with ROSS™ and ROSS Ultra™ pH electrodes to prevent damaging the ROSS electrodes
  • When the single junction reference electrode (900100) or double junction reference electrode (900200) are used with Orion ion selective electrodes (ISE), refer to the recommended ISE filling solution listed in the ISE user guide
Warranty: Out of box.
Specifications
Specifications
pH Electrode Filling Solution
60 mL
SDS
missing translation for 'documents'
missing translation for 'documents'
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.