missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oxalic acid, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 124280010

 View more versions of this product

Catalog No. AC124280010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
Specifications

Specifications

Oxalic acid
Colorless
HO2CCO2H
02, 502
ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico
MUBZPKHOEPUJKR-UHFFFAOYSA-N
0.0998 to 0.1002N (20°C)
90.034
CHEBI:16995
0.9900g/mL
Plastic bottle
0.99
144-62-7
C2H2O4
MFCD00002573
15, 7010
Solubility in water: soluble in water.
C(=O)(C(=O)O)O
oxalic acid
971
90.04
1 L
Liquid

Chemical Identifiers

144-62-7
90.034
MUBZPKHOEPUJKR-UHFFFAOYSA-N
971
oxalic acid
C2H2O4
MFCD00002573
ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico
CHEBI:16995
C(=O)(C(=O)O)O
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only