missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oxalic acid, Thermo Scientific Chemicals

$87.95 - $203.86

Chemical Identifiers

CAS 144-62-7
Molecular Formula C2H2O4
Molecular Weight (g/mol) 90.034
MDL Number MFCD00002573
InChI Key MUBZPKHOEPUJKR-UHFFFAOYSA-N
Synonym ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico
PubChem CID 971
ChEBI CHEBI:16995
IUPAC Name oxalic acid
SMILES C(=O)(C(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC124280010
SDS View Documents
Thermo Scientific Chemicals
124280010
1 L Plastic bottle
Each for $87.95
 
AC124280025
SDS View Documents
Thermo Scientific Chemicals
124280025
2.5 L Plastic bottle
Each for $203.86
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Volumetric standard solution
Specifications

Chemical Identifiers

144-62-7
90.034
MUBZPKHOEPUJKR-UHFFFAOYSA-N
971
oxalic acid
C2H2O4
MFCD00002573
ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico
CHEBI:16995
C(=O)(C(=O)O)O

Specifications

144-62-7
C2H2O4
MFCD00002573
15, 7010
Solubility in water: soluble in water.
C(=O)(C(=O)O)O
oxalic acid
971
90.04
1 L
Liquid
Oxalic acid
Colorless
HO2CCO2H
02, 502
ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico
MUBZPKHOEPUJKR-UHFFFAOYSA-N
0.0998 to 0.1002N (20°C)
90.034
CHEBI:16995
0.9900g/mL
Plastic bottle
0.99
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only