missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Oxalic acid, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 124280025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solutionSpecifications
Oxalic acid | |
Colorless | |
HO2CCO2H | |
02, 502 | |
ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico | |
MUBZPKHOEPUJKR-UHFFFAOYSA-N | |
0.0998 to 0.1002N (20°C) | |
90.034 | |
CHEBI:16995 | |
0.9900g/mL | |
Plastic bottle | |
0.99 |
144-62-7 | |
C2H2O4 | |
MFCD00002573 | |
15, 7010 | |
Solubility in water: soluble in water. | |
C(=O)(C(=O)O)O | |
oxalic acid | |
971 | |
90.04 | |
2.5 L | |
Liquid |
Chemical Identifiers
144-62-7 | |
90.034 | |
MUBZPKHOEPUJKR-UHFFFAOYSA-N | |
971 | |
oxalic acid |
C2H2O4 | |
MFCD00002573 | |
ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico | |
CHEBI:16995 | |
C(=O)(C(=O)O)O |
RUO – Research Use Only