Learn More
p-Cresol, 99+%, pure, Thermo Scientific Chemicals
$50.44 - $115.68
Chemical Identifiers
CAS | 106-44-5 |
---|---|
Molecular Formula | C7H8O |
Molecular Weight (g/mol) | 108.14 |
MDL Number | MFCD00002376 |
InChI Key | IWDCLRJOBJJRNH-UHFFFAOYSA-N |
Synonym | p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene |
PubChem CID | 2879 |
ChEBI | CHEBI:17847 |
IUPAC Name | 4-methylphenol |
SMILES | CC1=CC=C(C=C1)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AC405740050
|
Thermo Scientific Chemicals
405740050 |
5 g |
Each for $50.44
|
|
||||||
AC405741000
|
Thermo Scientific Chemicals
405741000 |
100 g |
Each for $75.80
|
|
||||||
AC405745000
|
Thermo Scientific Chemicals
405745000 |
500 g |
Each for $115.68
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
106-44-5 | |
108.14 | |
IWDCLRJOBJJRNH-UHFFFAOYSA-N | |
2879 | |
4-methylphenol |
C7H8O | |
MFCD00002376 | |
p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene | |
CHEBI:17847 | |
CC1=CC=C(C=C1)O |
Specifications
106-44-5 | |
Colorless to Yellow | |
202.0°C | |
Authentic | |
Glass bottle | |
CH3C6H4OH | |
5 g | |
1.034 | |
p-cresol, 4-cresol, 4-hydroxytoluene, p-methylphenol, phenol, 4-methyl, p-hydroxytoluene, p-tolyl alcohol, p-kresol, para-cresol, p-oxytoluene | |
IWDCLRJOBJJRNH-UHFFFAOYSA-N | |
4-methylphenol | |
2879 | |
CHEBI:17847 | |
99+% | |
Crystalline Solid or Liquid |
32.0°C to 34.0°C | |
1.0340g/mL | |
89°C | |
99% min. (GC) | |
C7H8O | |
MFCD00002376 | |
06,389 | |
15,2564 | |
Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol,ether and chloroform | |
CC1=CC=C(C=C1)O | |
108.14 | |
8 mPa.s (35°C) | |
108.14 | |
Pure | |
p-Cresol, 99+% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 203-398-6
RTECSNumber : GO6475000
TSCA : TSCA
RUO – Research Use Only