missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

p-Phenylenediamine dihydrochloride, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 417480250

 View more versions of this product

Catalog No. AC417480250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

p-Phenylenediamine dihydrochloride
624-18-0
Authentic
Glass bottle
C6H4(NH2)2·2HCl
25 g
Solubility in water: soluble.
C1=CC(=CC=C1N)N.Cl.Cl
181.06
181.06
Crystalline Powder
99+%
Gray-Purple to White
99% min. (Argentometry)
C6H8N2·2HCl
MFCD00013002
benzene-1,4-diamine dihydrochloride, p-phenylenediamine dihydrochloride, 1,4-diaminobenzene dihydrochloride, 1,4-phenylenediamine dihydrochloride, fourrine ds, 1,4-benzenediamine, dihydrochloride, pelagol cd, pelagol grey cd, durafur black rc, fourrine 64
IYXMNTLBLQNMLM-UHFFFAOYSA-N
benzene-1,4-diamine;dihydrochloride
12205
99+%

Chemical Identifiers

624-18-0
181.06
IYXMNTLBLQNMLM-UHFFFAOYSA-N
12205
C1=CC(=CC=C1N)N.Cl.Cl
C6H8N2·2HCl
MFCD00013002
benzene-1,4-diamine dihydrochloride, p-phenylenediamine dihydrochloride, 1,4-diaminobenzene dihydrochloride, 1,4-phenylenediamine dihydrochloride, fourrine ds, 1,4-benzenediamine, dihydrochloride, pelagol cd, pelagol grey cd, durafur black rc, fourrine 64
benzene-1,4-diamine;dihydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Toxic if swallowed.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-834-9

RTECSNumber : ST0350000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only